missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Chloride, Pellets, Purified, J.T. Baker™
769.50 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
Specifications
Physical Form | Pellets |
Quantity | 2.5 kg |
Formula Weight | 110.98 |
Grade | Purified |
Packaging | Poly Bottle |
Specific Gravity | 1L = 2.15kg |
Chemical Name or Material | Calcium Chloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'