missing translation for 'onlineSavingsMsg'
Learn More

Calcium Chloride Hexahydrate, 98.5%+, Extra Pure, SLR, Fisher Chemical™

Brand:  Fisher Chemical C/1250/53

Code : 21

Additional Details : CAS Number : 7774-34-7 Weight : 0.50000kg


 View more versions of this product

Product Code. 12638625

  • £69.95 / 250g

In stock, ready to ship: 2

Please to purchase this item. Need a web account? Register with us today!


You may only order up to the available stock. For more details call customer services on 01509 555 500

Description

Description

Specifications

Specifications

7774-34-7
149616
Solid
Colorless
6.5 to 8.0
CaCl2·6H2O
219.08g/mol
Plastic powder jar
30°C
500 g
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Calcium chloride hexahydrate, 98.5%+, Specified (SLR)

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS Documents