missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium chloride, Free-flowing, ACS reagent, anhydrous, ≥96%, Honeywell Fluka™
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Boiling Point | 1,900°C |
Melting Point | 772°C (literature) |
CAS Max % | 100.0000% |
Physical Form | Anhydrous |
Quantity | 500 g |
Linear Formula | CaCl2 |
Formula Weight | 110.98 |
Percent Purity | >96% |
Grade | ACS Reagent |
Packaging | Poly Bottle |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'