missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium chloride dihydrate, Puriss. p.a., ACS Reagent, Reag. Ph. Eur., ≥99%, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
50 kg
6 x 2.5 kg
Unit Size:
1kg
2.5kg
500g
50kg
Pack of 6
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Boiling Point | 1,900°C |
| Melting Point | 176°C (decomposition)(literature) |
| CAS Max % | 100.0000% |
| Quantity | 50 kg |
| Linear Formula | KSCN |
| MDL Number | MFCD00011413 |
| UN Number | NONH for all modes of transport |
| Formula Weight | 147.01 |
| Percent Purity | >99% |
| Grade | ACS Reagent/Reagent Ph. Eur. |
| Show More |
Safety and Handling
Product Identifier
- Calcium chloride dihydrate
- Warning
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction