missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Chloride Dihydrate, puriss., Honeywell Fluka™
Click to view available options
Quantity:
1 kg
2.5 kg
15 kg
6 x 1 kg
6 x 2.5 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Quantity | 6 x 1 kg |
UN Number | NONH for all modes of transport |
Formula Weight | 147.01 |
Percent Purity | 99 to 103% |
Grade | puriss. |
Chemical Name or Material | Calcium Chloride Dihydrate, Meets analytical specification of Ph. Eur.USP, FCC |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'