missing translation for 'onlineSavingsMsg'
Learn More

Calcium chloride dihydrate, 99% min, Thermo Scientific Chemicals

Catalog Number p-4412830
Click to view available options
Quantity:
500 g
2 kg
This item is not returnable. View return policy
10035-04-8
CaCl2·2H2O
MFCD00149613
This item is not returnable. View return policy
10035-04-8
CaCl2·2H2O
MFCD00149613

Calcium chloride dihydrate is widely used as an ingredient in ceramic slipware, cosmetic and food items. It acts as a pH buffer in swimming pool water. It provides inhibition of swelling clays in the water phase of invert emulsion drilling fluids. It is employed in the oil industry to increase the density of solids-free brines. It is used as a flux in the Davy process for the manufacture of sodium through the electrolysis of molten sodium chloride. It finds an important application as an additive in plastics, fire extinguishers, blast furnaces and in wastewater treatment as a drainage aid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Granules
Quantity 500 g
Assay Percent Range 99% min.
Solubility Information Soluble in water,acetic acid and alcohol.
Formula Weight 147.02 (110.99anhydrous)
Sensitivity Hygroscopic
Density 0.835 g/mL
Chemical Name or Material Calcium chloride dihydrate
compliance-icons
Product Identifier
  • Calcium chloride dihydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : calcium chloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'