missing translation for 'onlineSavingsMsg'
Learn More

Calcium chloride dihydrate, 99%, for biochemistry, Thermo Scientific Chemicals

Catalog Number 13525020
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Plastic Bottle
This item is not returnable. View return policy
10035-04-8
CaCl2·2H2O
147.02
This item is not returnable. View return policy
10035-04-8
CaCl2·2H2O
147.02

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 175.0°C
Quantity 2.5 kg
Assay Percent Range 98.5% min. (Complexometry)
Linear Formula CaCl2·2H2O
Merck Index 15,1661
Solubility Information Clear colorless
Formula Weight 147.02
Percent Purity 99%
Grade Biochemical
Packaging Plastic Bottle
Specific Gravity 0.83
Density 0.8300g/mL
Chemical Name or Material Calcium Chloride Dihydrate
Show More Show Less
compliance-icons
Product Identifier
  • Calcium chloride dihydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'