missing translation for 'onlineSavingsMsg'
Learn More

Calcium carbonate, Puratronic™, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog Number 11320919
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
This item is not returnable. View return policy
471-34-1
CCaO3
100.09
MFCD00010906
VTYYLEPIZMXCLO-UHFFFAOYSA-L
calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama
10112
CHEBI:3311
calcium;carbonate
[Ca++].[O-]C([O-])=O
This item is not returnable. View return policy
471-34-1
CCaO3
100.09
MFCD00010906
VTYYLEPIZMXCLO-UHFFFAOYSA-L
calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama
10112
CHEBI:3311
calcium;carbonate
[Ca++].[O-]C([O-])=O

Calcium carbonate is used in medicine and agriculture. It is also used in flue gas desulfurization in fossil fuel power stations, production of toothpaste, ceramics/glazing applications, purification of iron from iron ore, and to clean tarnish on silver. It forms the major component of ceramic tile adhesives. It is used in the refining of sugar, as filler in thermosetting resins, as an abrasive, as an extender in paints, and as a formation-bridging filtercake-sealing agent in oil industry. It is used in swimming pools for controlling the pH, to improve the rigidity of polypropylene compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 800°C (decomposition)
Color White
Physical Form Powder
Quantity 5 g
Merck Index 14,1657
Solubility Information Very low solubility in water.
Formula Weight 100.09
Percent Purity 99.99%
Grade Puratronic™
Odor Odorless
Refractive Index 1.6583
Density 2.930 g/mL
Assay (metals basis)
Chemical Name or Material Calcium carbonate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'