missing translation for 'onlineSavingsMsg'
Learn More

Calcium Carbonate, Light Powder, U.S.P. - F.C.C., J.T. Baker™

Catalog Number 15567894
356.25 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.


Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
Poly Pail
This item is not returnable. View return policy
471-34-1
CCaO3
100.09
MFCD00010906
VTYYLEPIZMXCLO-UHFFFAOYSA-L
calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama
10112
CHEBI:3311
[Ca++].[O-]C([O-])=O
This item is not returnable. View return policy
471-34-1
CCaO3
100.09
MFCD00010906
VTYYLEPIZMXCLO-UHFFFAOYSA-L
calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama
10112
CHEBI:3311
[Ca++].[O-]C([O-])=O
Physical Form Powder
Quantity 500 g
IUPAC Name calcium;carbonate
Formula Weight 100.09
Grade FCC/USP
Packaging Poly Bottle
Chemical Name or Material Calcium Carbonate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'