Learn More
Calcium carbonate, Puratronic™, 99.997% (metals basis), Thermo Scientific Chemicals
Use promo code "21615" to get your promotional price.
Description
Calcium carbonate is used in medicine and agriculture. It is also used in flue gas desulfurization in fossil fuel power stations, production of toothpaste, ceramics/glazing applications, purification of iron from iron ore, and to clean tarnish on silver. It is used in the refining of sugar, as filler in thermosetting resins, as an abrasive, as an extender in paints, and as a formation-bridging filtercake-sealing agent in oil industry. It is used in swimming pools for controlling the pH, to improve the rigidity of polypropylene compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 800°C (decomposition) |
Color | White |
Physical Form | Powder |
Quantity | 5 g |
Assay Percent Range | 99.997% (Metal basis) |
Solubility Information | Soluble in acids and insoluble in water. |
IUPAC Name | calcium carbonate |
Formula Weight | 100.09 |
Refractive Index | 1.6583 |
Appearance | White powder |
Show More |
RUO – Research Use Only