Learn More
Calcium boride, 99.5% (metals basis), Thermo Scientific Chemicals
Description
Calcium boride is used as an electrode (cathode) material and n-type thermoelectric materials. It is used to make boron- alloyed steel, as a deoxidation agent in the production of oxygen-free copper. It is further used as an antioxidant in carbon bonded refractories. It acts as a new material which is used in the nuclear-industry for neutron- preventing, high-purity metal borid (TiB2,ZrB2,HfB2) and high-purity boron alloy(Ni-B, Co-B, Cu-b).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 2235°C |
Physical Form | Powder |
Quantity | 5 g |
Solubility Information | Insoluble in water. |
Formula Weight | 104.95 |
Percent Purity | 99.5% |
Density | 2.30 g/mL |
Chemical Name or Material | Calcium boride |
RUO – Research Use Only