missing translation for 'onlineSavingsMsg'
Learn More

Calcium 2,4-pentanedionate hydrate, 99%, Thermo Scientific Chemicals

Catalog Number p-7049873
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
345909-31-1
C10H14CaO4
238.30
MFCD00013486 MFCD00151667
QAZYYQMPRQKMAC-FDGPNNRMSA-L
calcium bis 4-oxopent-2-en-2-olate hydrate
16219078
[Ca++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
This item is not returnable. View return policy
345909-31-1
C10H14CaO4
238.30
MFCD00013486 MFCD00151667
QAZYYQMPRQKMAC-FDGPNNRMSA-L
calcium bis 4-oxopent-2-en-2-olate hydrate
16219078
[Ca++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 175°C (decomposition)
Quantity 10 g
IUPAC Name calcium bis((2Z)-4-oxopent-2-en-2-olate)
Formula Weight 238.30 (Anhydrous)
Percent Purity 99%
Physical Form Crystalline
Chemical Name or Material Calcium 2,4-pentanedionate hydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'