missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Calcium 2,4-pentanedionate hydrate, 99%

Brand:  Alfa Aesar™ 030526.18

Code : NEW

Additional Details : CAS Number : 345909-31-1 Weight : 0.05000kg

Quantity 50g

 View more versions of this product

Product Code. 11317298

  • £39.70 / 50g


Estimated Shipment date
from Supplier 02-02-2018
Add to basket

Specifications

Chemical Identifiers

Calcium 2,4-pentanedionate hydrate
238.30 (Anhydrous)
Calcium acetylacetonate
345909-31-1
MFCD00013486

Specifications

Calcium 2,4-pentanedionate hydrate
Crystalline
238.30 (Anhydrous)
Calcium acetylacetonate
50g
345909-31-1
99%
MFCD00013486
175°C (dec.)
Videos
SDS