Learn More
Calciferol, 97%, Thermo Scientific Chemicals
Description
Calciferol has a strong inhibitory effect against bladder tumor promotion by sodium saccharin and induces cell differentiation in leukemia cells. Calciferol has also been shown to be an inhibitor of DNA polymerase. Calciferol with calcium is used to treat or prevent bone loss (osteoporosis). Calciferol is also used with other medications to treat low levels of calcium or phosphate caused by certain disorders (such as hypoparathyroidism, pseudohypoparathyroidism, familial hypophosphatemia). It is often used in kidney disease to keep calcium levels normal and allow normal bone growth.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Soluble in ethanol (80 mg/ml at 25°C), water (<1mg/mL at 25°C), methanol, DMF (∼20 mg/ml), and DMSO (20 mg/ml at 25°C).
Notes
Store at 4°C. Incompatible with heat and oxidizing agents.
Specifications
Melting Point | 118°C to 119°C |
Color | White |
Quantity | 1 g |
UN Number | UN2811 |
Beilstein | 1916682 |
Merck Index | 14,10018 |
Solubility Information | Soluble in ethanol (80mg/ml at 25°C),water (<1mg/ml at 25°C),methanol,DMF (∽20mg/ml),and DMSO (20mg/ml at 25°C). |
Optical Rotation | +110° (c=1.5 in Ethanol) |
Formula Weight | 396.66 |
Percent Purity | 97% |
Show More |
RUO – Research Use Only