missing translation for 'onlineSavingsMsg'
Learn More

Merck Millipore Calbiochem™ EMPIGEN™ BB Detergent, 30% Solution

Can be used as detergent for solubilization of membrane components

Brand:  Merck Millipore 324690-100ML

Additional Details : Weight : 0.10000kg Transport : UN number : 2810 Chem class : 6.1 Pack group : III

Product Code. 15890925

  • £133.90 / 100mL

Estimated Shipment
19-04-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Zwitterionic detergent
  • Useful in preparation of viral antigens, solubilization of keratins
Specifications

Specifications

100mL
15° to 30°C
BB Detergent, 30% Solution
Videos
SDS
Documents

For Research Use Only