missing translation for 'onlineSavingsMsg'
Learn More

Caffeine, 99%, Thermo Scientific Chemicals

Catalog Number p-7080758
57.05 GBP valid until 2024-04-30
Use promo code "21741" to get your promotional price.


Click to view available options
Quantity:
100 g
250 g
1000 g
58-08-2
C8H10N4O2
194.19
MFCD00005758
RYYVLZVUVIJVGH-UHFFFAOYSA-N
caffeine, 1,3,7-trimethylxanthine, guaranine, thein, cafeina, methyltheobromine, koffein, mateina, theine, alert-pep
2519
CHEBI:27732
CN1C=NC2=C1C(=O)N(C)C(=O)N2C
58-08-2
C8H10N4O2
194.19
MFCD00005758
RYYVLZVUVIJVGH-UHFFFAOYSA-N
caffeine, 1,3,7-trimethylxanthine, guaranine, thein, cafeina, methyltheobromine, koffein, mateina, theine, alert-pep
2519
CHEBI:27732
CN1C=NC2=C1C(=O)N(C)C(=O)N2C

This material is traded for research purposes ONLY. Thermo Fisher does NOT certify this material as suitable for pharmaceutical, cosmetic, veterinary or food use. Caffeine has inhibitory action against lung and colon tumorigenses and virus induction.Caffeine is used to treat and prevent bronchopulmonary dysplasia in premature infants. It is also used for the treatment of apnea of prematurity and orthostatic hypotension. It is widely used as an additive in food, cosmetics, pharmaceuticals and beverages. It confers a protective effect against some diseases including Parkinson's disease and certain types of cancer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in pyrrole, tetrahydrofuran+ water mixture and ethyl acetate. Slightly soluble in water and ethanol.

Notes
Store in cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 234°C to 236°C
Density 1.23
Assay Percent Range 99%
Quantity 100 g
UN Number UN1544
Beilstein 17705
Merck Index 14,1636
Solubility Information Soluble in pyrrole,tetrahydrofuran+water mixture and ethyl acetate; Slightly soluble in water and ethanol.
IUPAC Name 1,3,7-trimethylpurine-2,6-dione
Formula Weight 194.19
Percent Purity 99%
Chemical Name or Material Caffeine
Show More Show Less
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'