Learn More
Caffeic acid phenethyl ester, Tocris Bioscience™
Specific inhibitor of NF-κB activation
Brand: Tocris (Bio-Techne) 2743/10
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity | 10mg |
---|
Description
Antioxidant, antimitogenic, anticarcinogenic, anti-inflammatory and antiviral. Specifically inhibits NF-κB activation and inhibits the growth of transformed cells. Also suppresses lipid peroxidation and inhibits ornithine decarboxylase, protein tyrosine kinase and lipoxygenase activities.Chemical Identifiers
104594-70-9 | |
284.311 | |
caffeic acid phenethyl ester, phenethyl caffeate, cape, capeee, phenethyl 3-3,4-dihydroxyphenyl acrylate, phenylethyl caffeate, unii-g960r9s5sk, caffeic acid phenylethyl ester, 2-phenylethyl caffeate, caffeic acid 2-phenylethyl ester | |
CHEBI:8062 | |
C1=CC=C(C=C1)CCOC(=O)C=CC2=CC(=C(C=C2)O)O |
C17H16O4 | |
SWUARLUWKZWEBQ-VQHVLOKHSA-N | |
5281787 | |
2-phenylethyl (E)-3-(3,4-dihydroxyphenyl)prop-2-enoate |
Specifications
Caffeic acid phenethyl ester | |
104594-70-9 | |
caffeic acid phenethyl ester, phenethyl caffeate, cape, capeee, phenethyl 3-3,4-dihydroxyphenyl acrylate, phenylethyl caffeate, unii-g960r9s5sk, caffeic acid phenylethyl ester, 2-phenylethyl caffeate, caffeic acid 2-phenylethyl ester | |
C1=CC=C(C=C1)CCOC(=O)C=CC2=CC(=C(C=C2)O)O | |
284.311 | |
CHEBI:8062 | |
>99% |
10mg | |
C17H16O4 | |
SWUARLUWKZWEBQ-VQHVLOKHSA-N | |
2-phenylethyl (E)-3-(3,4-dihydroxyphenyl)prop-2-enoate | |
5281787 | |
284.31 |