missing translation for 'onlineSavingsMsg'
Learn More

Cadmium telluride, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog Number 15402277
Click to view available options
Quantity:
1 g
25 g
Packaging:
Glass bottle
Plastic bottle
This item is not returnable. View return policy
1306-25-8
CdTe
240.01
MFCD00015998
RPPBZEBXAAZZJH-UHFFFAOYSA-N
cadmium telluride, irtran 6, cadmium monotelluride, unii-stg188wo13, cadmiumtellurid, irtran-6, cadmium telluride metals basis, cadmium telluride trace metals basis
91501
tellanylidenecadmium
[Cd]=[Te]
This item is not returnable. View return policy
1306-25-8
CdTe
240.01
MFCD00015998
RPPBZEBXAAZZJH-UHFFFAOYSA-N
cadmium telluride, irtran 6, cadmium monotelluride, unii-stg188wo13, cadmiumtellurid, irtran-6, cadmium telluride metals basis, cadmium telluride trace metals basis
91501
tellanylidenecadmium
[Cd]=[Te]

Used to make thin film solar cells.

Cadmium telluride is used to make thin film solar cells. It can be alloyed with mercury to make a versatile infrared detector material (HgCdTe). It is used as an infrared optical material for optical windows and lenses and is proven to provide a good performance across a wide range of temperatures. It is also applied for electro-optic modulators and has the greatest electro-optic coefficient of the linear electro-optic effect among II-VI compound crystals. It can be doped with chlorine, which is used as a radiation detector for x-rays, gamma rays, beta particles and alpha particles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 1041°C
Physical Form Polycrystalline Pieces
Quantity 1 g
UN Number UN3077
Solubility Information Insoluble in water.
Formula Weight 240
Percent Purity 99.999%
Packaging Plastic bottle
Specific Gravity 6.2
Density 6.2
Chemical Name or Material Cadmium telluride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'