missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cadmium sulfate octahydrate, ACS, 98.0-102.0%, Thermo Scientific Chemicals
46.60 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
25 g
250 g
1 kg
Description
Cadmium sulfate mainly used in electroplating of cadmium, which is used in electronic circuits. It is used as a precursor for cadmium based pigments such as cadmium sulfide and pharmaceutical intermediates. It is used in the standard weston cell.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | >40°C (in -H2O) |
Color | White |
Physical Form | Crystalline |
Quantity | 250 g |
Molecular Formula | 3CdSO4·8H2O |
UN Number | UN2570 |
Merck Index | 14,1627 |
Solubility Information | Highly soluble in water; Partially soluble in methanol and ethyl acetate; Insoluble in ethanol. |
Formula Weight | 769.50 (208.46 Anhydrous) |
Percent Purity | 98.0 to 102.0% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'