Cadmium Sulfate, Hydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

Overview
Brand: JT Baker

Manufacturer Part Number: 1243-04

UNSPSC: 12000000

Additional Details:
Additional Details: CAS Number: 7790-84-3 Weight: 0.62000kg


Packaging Wide Mouth Amber Glass Bottle
Quantity 125g

Product Code. 15547894

Quantity Price
1 £ 167.2 / 125g
Call for latest availability
+44 (0)1509 555 500
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material Cadmium Sulfate
CAS 7790-84-3
Name Note Hydrate
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula CdSO4.8/3 H2O
Formula Weight 256.52

Specification

Purity Grade Baker Analyzed™ ACS Reagent
Physical Form Crystals
Formula Weight 256.52
Chemical Name or Material Cadmium Sulfate
CAS 7790-84-3
Molecular Formula CdSO4.8/3 H2O
Specific Gravity 1L = 3.08kg
Name Note Hydrate
Packaging Wide Mouth Amber Glass Bottle
Quantity 125g