missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cadmium Sulfate 8/3-Hydrate, Honeywell Fluka™
Click to view available options
Quantity:
500 g
1 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Quantity | 1 kg |
Linear Formula | CdSO4 · 8/3H2O |
UN Number | 2570 |
Formula Weight | 256.57g/mol |
Percent Purity | ≥98% (complexometric) |
Grade | puriss. |
Chemical Name or Material | Cadmium Sulfate 8/3-Hydrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'