missing translation for 'onlineSavingsMsg'
Learn More

Cadmium iodide, ultra dry, 99.9985% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7050130
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
7790-80-9
CdI2
366.223
MFCD00010920
OKIIEJOIXGHUKX-UHFFFAOYSA-L
cadmium iodide, cadmium diiodide, unii-2f2upu4kcw, cadmium 2+ diiodide, 2f2upu4kcw, acmc-20aljc, cadmium 2+ ion diiodide, cadmium iodide trace metals basis 25g
24635
cadmium(2+);diiodide
[Cd+2].[I-].[I-]
This item is not returnable. View return policy
7790-80-9
CdI2
366.223
MFCD00010920
OKIIEJOIXGHUKX-UHFFFAOYSA-L
cadmium iodide, cadmium diiodide, unii-2f2upu4kcw, cadmium 2+ diiodide, 2f2upu4kcw, acmc-20aljc, cadmium 2+ ion diiodide, cadmium iodide trace metals basis 25g
24635
cadmium(2+);diiodide
[Cd+2].[I-].[I-]

Cadmium iodide is used in lithography, photography, electroplating and the manufacturing of phosphors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY
Boiling Point 787°C
Melting Point 387°C to 389°C
Color White to Yellow
Physical Form Beads
Quantity 1 g
UN Number UN2570
Merck Index 14,1621
Solubility Information It is soluble in water,ethanol,acetone,ether and ammonia.
Formula Weight 366.21
Percent Purity 99.9985%
Odor Odorless
Packaging Ampule
Sensitivity Hygroscopic; Light sensitive
Density 5.67 g/mL
Assay (metals basis)
Chemical Name or Material Cadmium iodide, Ultra dry
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'