missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cadmium iodide, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | White to Yellow |
Physical Form | Shiny Crystalline Flakes or Powder |
Quantity | 250 g |
Assay Percent Range | 99% |
Molecular Formula | CdI2 |
Linear Formula | CdI2 |
Merck Index | 15, 1626 |
Molecular Weight (g/mol) | 366.223 |
Formula Weight | 366.2 |
Percent Purity | ≥98.5% |
Show More |
Safety and Handling
Product Identifier
- Cadmium iodide
- Danger
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Carcinogenicity Category 2
- Specific target organ toxicity after repeated exposure Category 2
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H351-Suspected of causing cancer.
- H373-May cause damage to organs.
- H331-Toxic if inhaled.
- H301-Toxic if swallowed.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'