missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Cadmium chloride, ultra dry, 99.9% (metals basis)

Brand:  Alfa Aesar™ 035728.09

Code : NEW

Additional Details : CAS Number : 10108-64-2 Weight : 0.01000kg Transport : UN number : 2570 Chem class : 1 Pack group : III

Quantity 10g

 View more versions of this product

Product Code. 11318597

  • £88.51 / 10g


Estimated Shipment date
from Supplier 19-02-2018
Add to basket

Specifications

Chemical Identifiers

Cadmium chloride
Ultra dry
MFCD00010916
14,1617
10108-64-2
183.32
UN2570

Specifications

Cadmium chloride
10108-64-2
99.9%
183.32
UN2570
4.047g/mL
Odorless
Hygroscopic
568°C
Ultra dry
Beads
(metals basis)
MFCD00010916
14,1617
−10 Mesh
Ampule
960°C
10g
Videos
SDS