missing translation for 'onlineSavingsMsg'
Learn More

Cadmium chloride hydrate, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7048481
Click to view available options
Quantity:
1 g
10 g
50 g
This item is not returnable. View return policy
654054-66-7
CdCl2
183.31
MFCD00149627
YKYOUMDCQGMQQO-UHFFFAOYSA-L
cadmium chloride, ultra dry, cdcl2.h2o, cadmium chloride hydrate, cadmium chloride 1-hydrate, cadmium chloride hydrate, puratronic, cadmium chloride monohydrate, puriss, cadmium chloride hydrate trace metals basis
16211517
[Cl-].[Cl-].[Cd++]
This item is not returnable. View return policy
654054-66-7
CdCl2
183.31
MFCD00149627
YKYOUMDCQGMQQO-UHFFFAOYSA-L
cadmium chloride, ultra dry, cdcl2.h2o, cadmium chloride hydrate, cadmium chloride 1-hydrate, cadmium chloride hydrate, puratronic, cadmium chloride monohydrate, puriss, cadmium chloride hydrate trace metals basis
16211517
[Cl-].[Cl-].[Cd++]

Cadmium chloride hydrate is used in dyeing, printing, textiles and in metal finishing baths. It is also used as a reagent to make photographic paper and carbon paper. Further, it is utilized as an absorbing agent to absorb hydrogen sulfide as well as analytical reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Crystalline Powder
Quantity 1 g
UN Number UN2570
Merck Index 14,1617
IUPAC Name dichlorocadmium;hydrate
Formula Weight 183.32 (Anhydrous)
Percent Purity 99.998%
Grade Puratronic™
Refractive Index 1.6513
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Cadmium chloride hydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'