missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cadmium Chloride, Honeywell™
Click to view available options
Quantity:
25 g
100 g
500 g
Description
- We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
Specifications
Physical Form | Anhydrous |
Quantity | 100 g |
UN Number | 2570 |
Solubility Information | Soluble in water 457g/L at 20°C |
Formula Weight | 183.32g/mol |
Percent Purity | ≥99% |
Grade | puriss. p.a. |
Vapor Pressure | 10mmHg (656°C) |
Density | Vapor Density: 6.3 (vs air) |
Chemical Name or Material | Cadmium Chloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'