missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cadmium chloride hemipentahydrate, ACS, 79.5-81.0%, Thermo Scientific Chemicals
22.33 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
50 g
250 g
Description
Cadmium chloride is also used for photocopying, dyeing and electroplating. Useful in pyrotechnic devices.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | White |
Physical Form | Crystalline |
Quantity | 50 g |
Assay Percent Range | 79.5-81.0% |
Molecular Formula | CdCl2·2.5H2O |
Solubility Information | Highly soluble in water and slightly soluble in alcohol. |
Formula Weight | 228.36 (183.32 Anhydrous) |
Grade | ACS |
Odor | Odorless |
Refractive Index | 1.6513 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'