missing translation for 'onlineSavingsMsg'
Learn More

Cadmium carbonate, >99% (metals basis), Thermo Scientific Chemicals

Catalog Number 11371579
Click to view available options
Quantity:
250 g
1 kg
This item is not returnable. View return policy
513-78-0
CCdO3
172.42
MFCD00010918
GKDXQAKPHKQZSC-UHFFFAOYSA-L
cadmium carbonate, mikrokalcit, otavite, kalcit, supermikrokalcit, carbonic acid, cadmium salt 1:1, cadmium monocarbonate, chemcarb cdco3, caswell no. 134a, unii-616zvs3bly
10564
cadmium(2+);carbonate
[Cd++].[O-]C([O-])=O
This item is not returnable. View return policy
513-78-0
CCdO3
172.42
MFCD00010918
GKDXQAKPHKQZSC-UHFFFAOYSA-L
cadmium carbonate, mikrokalcit, otavite, kalcit, supermikrokalcit, carbonic acid, cadmium salt 1:1, cadmium monocarbonate, chemcarb cdco3, caswell no. 134a, unii-616zvs3bly
10564
cadmium(2+);carbonate
[Cd++].[O-]C([O-])=O

Cadmium carbonate is the raw material for the production of pigments for ceramic and frits. Used as reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 500°C (decomposition)
Color White
Physical Form Powder
Quantity 250 g
Assay Percent Range >99% (Metal basis)
Solubility Information Very slightly soluble in water. Soluble in dilute acids,concentrated solutions of ammonium salts.
Formula Weight 172.42
Odor Odorless
Appearance White powder
Density 4.26 g/cm3
Chemical Name or Material Cadmium carbonate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'