missing translation for 'onlineSavingsMsg'
Learn More

Cadmium bromide hydrate, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7059387
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
681435-25-6
272.22
MFCD00150204
KPWJBEFBFLRCLH-UHFFFAOYSA-L
cadmium 2+ dibromide, cadmium 2+ ion dibromide, cadmium bromide, anhydrous 25g, cadmium bromide, anhydrous,
cadmium(2+) dibromide
[Br-].[Br-].[Cd++]
This item is not returnable. View return policy
681435-25-6
272.22
MFCD00150204
KPWJBEFBFLRCLH-UHFFFAOYSA-L
cadmium 2+ dibromide, cadmium 2+ ion dibromide, cadmium bromide, anhydrous 25g, cadmium bromide, anhydrous,
cadmium(2+) dibromide
[Br-].[Br-].[Cd++]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White to Yellow
Physical Form Crystalline
Quantity 25 g
Molecular Formula Br2Cd
UN Number UN2570
Formula Weight 272.22 (Anhydrous)
Percent Purity 99.999%
Odor Odorless
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Cadmium bromide hydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'