missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cadmium bromide, anhydrous, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
Cadmium bromide is used in process engraving, lithography and photography.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | 863°C |
Melting Point | 567°C |
Color | White to Yellow |
Physical Form | Powder |
Quantity | 25 g |
Assay Percent Range | 98.00% |
Solubility Information | Freely soluble in water,alcohol. Slightly soluble in acetone and ether. |
Formula Weight | 272.22 |
Odor | Odorless |
Sensitivity | Hygroscopic |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'