missing translation for 'onlineSavingsMsg'
Learn More

Cadmium acetate dihydrate, 98%, Thermo Scientific Chemicals

Catalog Number p-7051579
Click to view available options
Quantity:
100 g
500 g
2 kg
This item is not returnable. View return policy
5743-04-4
C4H10CdO6
266.53
MFCD00012449
AUIZLSZEDUYGDE-UHFFFAOYSA-L
This item is not returnable. View return policy
5743-04-4
C4H10CdO6
266.53
MFCD00012449
AUIZLSZEDUYGDE-UHFFFAOYSA-L

In cadium electroplating

Cadmium acetate dihydrate is in the porcelain production for the production of iridescent used effects. Moreover, it is in the analytical chemistry, a reagent for the detection of sulfur, selenium and tellurium. It is also used as chemical and pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Physical Form Powder
Quantity 100 g
UN Number UN2570
Merck Index 14,1614
Solubility Information Freely soluble in water, soluble in alcohol
Formula Weight 266.54 (230.50 Anhydrous)
Percent Purity 98%
Sensitivity Hygroscopic
Density 2.01 g/mL
Chemical Name or Material Cadmium acetate dihydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'