missing translation for 'onlineSavingsMsg'
Learn More

Cabergoline, Tocris Bioscience™

Selective D2-like agonist

Brand:  Tocris (Bio-Techne) 266450

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13431684

  • £576.24 / 50mg

Estimated Shipment date
from Supplier 27-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Selective D2-like dopamine receptor agonist (Ki values are 0.7, 1.5, 9.0 and 165 nM for D2, D3, D4 and D5 receptors respectively) that also displays high affinity for several serotonin receptor subtypes (Ki = 1.2 - 20.0 nM for 5-HT1A, 5-HT1D, 5-HT2A and 5-HT2B). Inhibits secretion of prolactin and growth hormone and reverses levodopa-induced dyskinesias in Parkinsonian monkeys.
Specifications

Chemical Identifiers

81409-90-7
451.615
cabergoline, dostinex, cabaser, cabergolina, cabergolinum, cabaseril, cabergolinum latin, cabergolina spanish, galastop, sogilen
CHEBI:3286
CCNC(=O)N(CCCN(C)C)C(=O)C1CC2C(CC3=CNC4=CC=CC2=C34)N(C1)CC=C
C26H37N5O2
KORNTPPJEAJQIU-KJXAQDMKSA-N
54746
(6aR,9R,10aR)-N-[3-(dimethylamino)propyl]-N-(ethylcarbamoyl)-7-prop-2-enyl-6,6a,8,9,10,10a-hexahydro-4H-indolo[4,3-fg]quinoline-9-carboxamide

Specifications

50mg
81409-90-7
cabergoline, dostinex, cabaser, cabergolina, cabergolinum, cabaseril, cabergolinum latin, cabergolina spanish, galastop, sogilen
CCNC(=O)N(CCCN(C)C)C(=O)C1CC2C(CC3=CNC4=CC=CC2=C34)N(C1)CC=C
451.615
CHEBI:3286
>99%
Cabergoline
C26H37N5O2
KORNTPPJEAJQIU-KJXAQDMKSA-N
(6aR,9R,10aR)-N-[3-(dimethylamino)propyl]-N-(ethylcarbamoyl)-7-prop-2-enyl-6,6a,8,9,10,10a-hexahydro-4H-indolo[4,3-fg]quinoline-9-carboxamide
54746
451.6
Videos
Documents