missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CAB39L Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA561134
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: MPLFSKSHKN PAEIVKILKD NLAI Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
CAB39L Polyclonal antibody specifically detects CAB39L in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Specifications
CAB39L | |
Polyclonal | |
Unconjugated | |
CAB39L | |
1500031K13Rik; 2810425O13Rik; 4930520C08Rik; AA589432; antigen MLAA-34; bA103J18.3; CAB39L; calcium binding protein 39 like; calcium binding protein 39-like; calcium-binding protein 39-like; MO25beta; MO25-BETA; mo25-like protein; MO2L; RP11-103J18.3; sarcoma antigen NY-SAR-79; U937-associated antigen | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
81617 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H9S4 | |
CAB39L | |
Recombinant protein corresponding to Human CAB39L | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only