missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CAB39L Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA550365
Additional Details : Weight : 0.01000kg
Description
The antibody detects endogenous levels of total CAB39L protein.
CAB39L Polyclonal antibody specifically detects CAB39L in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
CAB39L | |
Polyclonal | |
Unconjugated | |
CAB39L | |
1500031K13Rik; 2810425O13Rik; 4930520C08Rik; AA589432; antigen MLAA-34; bA103J18.3; CAB39L; calcium binding protein 39 like; calcium binding protein 39-like; calcium-binding protein 39-like; MO25beta; MO25-BETA; mo25-like protein; MO2L; RP11-103J18.3; sarcoma antigen NY-SAR-79; U937-associated antigen | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
81617 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1.6 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q9H9S4 | |
CAB39L | |
Fusion protein corresponding to residues near the C terminal of human calcium binding protein 39-like | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |