missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C1orf162 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA555474
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: NGSTCKPDTE RQGTLSTAAP TTSPAPCLSN HHNK Highest antigen sequence identity to the following orthologs: Mouse - 37%, Rat - 38%.
C1orf162 Polyclonal antibody specifically detects C1orf162 in Human samples. It is validated for Immunohistochemistry.Specifications
C1orf162 | |
Polyclonal | |
Unconjugated | |
C1orf162 | |
C1orf162; chromosome 1 open reading frame 162; transmembrane protein C1orf162 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
128346 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NEQ5 | |
C1orf162 | |
Recombinant protein corresponding to Human C1orf162 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only