Learn More
C-15 through 2-35 Odd Alkanes, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Brand: SPEX Certiprep ENC-ODD-1K
Code : NEW
Additional Details : Weight : 0.00100kg Transport : UN number : 1593 Chem class : 6.1 Pack group : III
Includes: Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
Analyte | CAS No. | Concentration |
---|---|---|
n-Heneicosane | 629-94-7 | 1000μg/mL |
n-Hentriacontane | 630-04-6 | 1000μg/mL |
n-Heptacosane | 593-49-7 | 1000μg/mL |
n-Heptadecane | 629-78-7 | 1000μg/mL |
n-Nonacosane | 630-03-5 | 1000μg/mL |
n-Nonadecane | 629-92-5 | 1000μg/mL |
n-Pentacosane | 629-99-2 | 1000μg/mL |
n-Pentadecane | 629-62-9 | 1000μg/mL |
n-Pentatriacontane | 630-07-9 | 1000μg/mL |
n-Tricosane | 638-67-5 | 1000μg/mL |
n-Tritriacontane | 630-05-7 | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
C-15 through 2-35 Odd Alkanes | |
Pentadecane; Heptadecane; Nonadecane; Heneicosane; Tricosane; Pentacosane; Heptacosane; Nonacosane; Hentriacontane; Tritricontane; Pentatriacontane | |
1 mL | |
Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity. |
2mL amber prescored glass ampule | |
MeCl | |
GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation |
Safety and Handling
- C15 THROUGH C35 ALK MIX
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Flammable liquid Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H351-Suspected of causing cancer.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : dichloromethane
Product intended for laboratory use only