missing translation for 'onlineSavingsMsg'
Learn More

c-Rel Rat anti-Mouse, eFluor 660, Clone: 1RELAH5, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 50-6111-82

Code : NEW

Additional Details : Weight : 0.09500kg

Product Code. 15569996

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Description

Description

The 1RELAH5 monoclonal antibody reacts with mouse c-Rel. c-Rel is a 65kDa hematopoietic lineage-restricted member of the NF-kB family of transcription factors. In the thymus, c-Rel is upregulated in regulatory T cells (Treg) when compared to other CD4+ thymocytes. c-Rel is believed to play a critical role in thymic Treg (nTreg) development, but is not necessary for TGF beta-mediated T reg (iTreg) differentiation. C-Rel is also involved in proliferation and survival in mature T cells.
Specifications

Specifications

c-Rel
1RELAH5
eFluor 660
Conjugated
Rat
100μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
0.2mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Rel
IgG2a, kappa
RUO
Primary
Murine
Videos
Documents

For Research Use Only.