missing translation for 'onlineSavingsMsg'
Learn More

c-Met, clone: eBioclone 7, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-8854-80

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15288097

  • £55.67 / 25µg


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

Description: The eBioclone 7 monoclonal antibody was generated against a mouse c-Met-Ig fusion protein, and reacts with mouse c-Met-transfected cells. Mouse c-Met (HGFR) is a 170kDa receptor tyrosine kinase (RTK) expressed by epithelial cells of the brain, kidney, liver and other tissues. Binding of its ligand, Hepatocyte Growth Factor (HGF), triggers receptor autophosphorylation, and activation of several downstream effectors including the mitogen-activated protein kinases ERK-1 and ERK-2, and PLC gamma. Activation of the c-Met signal transduction pathway leads to mulitple cellular responses including cell motility, scattering, proliferation, survival and angiogenesis. Mutations in human c-Met have been implicated in the development of several malignancies.Applications Reported: This eBioclone 7 antibody has been reported for use in flow cytometric analysis.Applications Tested: This eBioclone 7 antibody has been tested by flow cytometric analysis of mouse c-Met-transfected 293T cells. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

c-Met (HGF Receptor)
Monoclonal
0.5mg/mL
PBS with 0.09% sodium azide; pH 7.2
P16056
AUTS9, HGFR, RCCP2, c-Met
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
eBioclone 7
Unconjugated
Met
Purified
Met
IgG1, kappa
25μg
Store at 2-8°C.
Primary
17295
Videos

For Research Use Only.