missing translation for 'onlineSavingsMsg'
Learn More

IKA™ C 723 Benzoic Acid

Contains 0.5g per pellet and sealed in a blister package. IKA™ C723 Benzoic Acid includes IKA-certified gross calorific value for standard calibrations according to DIN 51900.

Brand:  IKA™ 0003243000

Code : A0

Additional Details : CAS Number : 65-85-0 Weight : 0.02500kg


Packaging Blister
Quantity 0.5g

Product Code. 10222401

  • £176.00 / Pack of 50

Estimated Shipment
17-11-2020

Please to purchase this item. Need a web account? Register with us today!



Description

Description

  • 0.5 g per pellet
  • Pelleted, blister package;
  • IKA-certified gross calorific value for standard calibrations according to DIN 51900
Specifications

Specifications

Blister
0.5g
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • benzoic acid

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 1

Hazard Statement
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H372-Causes damage to organs.
  • H315-Causes skin irritation.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.

Supplemental information
  • MIXTURE LIST-Contains : benzoic acid

Documents