missing translation for 'onlineSavingsMsg'
Learn More

C-1, Tocris Bioscience™

Protein kinase C inhibitor

Brand:  Tocris (Bio-Techne) 054310

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13447703

  • £127.89 / 10mg

Estimated Shipment date
from Supplier 20-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Protein kinase inhibitor. Inhibits PKG, PKA and PKC (IC50 values are 4, 8 and 12 ?M, respectively). Widely used as a ROCK inhibitor. Increases efficiency of human fibroblast reprogramming with PD 0325901 (Cat.No 4192), CHIR 99021 (Cat.No 4432), A 83-01 (Cat.No 2939) and hLIF. Improves survival of human pluripotent stem cells in culture.
Specifications

Chemical Identifiers

84468-24-6
277.342
1-5-isoquinolinesulfonyl piperazine, ha 100 pharmaceutical, 1-5-isoquinolinylsulfonyl piperazine, chembl75773, 5-piperazine-1-sulfonyl isoquinoline, piperazine, 1-5-isoquinolinylsulfonyl, iqsp, 5-piperazin-1-ylsulfonyl isoquinoline, tocris-0543, c-i-qp
5-piperazin-1-ylsulfonylisoquinoline
C13H15N3O2S
UPTYCYWTFGTCCG-UHFFFAOYSA-N
3545
C1CN(CCN1)S(=O)(=O)C2=CC=CC3=C2C=CN=C3

Specifications

10mg
84468-24-6
1-5-isoquinolinesulfonyl piperazine, ha 100 pharmaceutical, 1-5-isoquinolinylsulfonyl piperazine, chembl75773, 5-piperazine-1-sulfonyl isoquinoline, piperazine, 1-5-isoquinolinylsulfonyl, iqsp, 5-piperazin-1-ylsulfonyl isoquinoline, tocris-0543, c-i-qp
C1CN(CCN1)S(=O)(=O)C2=CC=CC3=C2C=CN=C3
277.342
277.34
C-1
C13H15N3O2S
UPTYCYWTFGTCCG-UHFFFAOYSA-N
5-piperazin-1-ylsulfonylisoquinoline
3545
>99%
Videos
Documents