Learn More
Tocris Bioscience™ BYK 204165
Selective PARP-1 inhibitor
Brand:  Tocris Bioscience™ 373410
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13463514
  • £124.20 / 10mg
Estimated Shipment
03-04-2019
Add to basket
Description
Description
Potent and selective poly(ADP-ribose) polymerase (PARP)-1 inhibitor (pIC 50 values are 5.38 and 7.35 for PARP-2 and PARP-1 respectively).
Specifications
Specifications
BYK 204165
>99
10mg
BYK 204165
252.27