Learn More
Tocris Bioscience™ BYK 204165
Selective PARP-1 inhibitor
POA
Specifications
Purity >99%
View More Specs
Description
Description
Potent and selective poly(ADP-ribose) polymerase (PARP)-1 inhibitor (pIC50 values are 5.38 and 7.35 for PARP-2 and PARP-1 respectively).
Specifications
Specifications
>99%