missing translation for 'onlineSavingsMsg'
Learn More

BW-B 70C, Tocris Bioscience™

5-Lipoxygenase inhibitor

Brand:  Tocris (Bio-Techne) 130410

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13429093

  • £139.65 / 10mg

Estimated Shipment date
from Supplier 27-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent and selective inhibitor of 5-lipoxygenase. Long half-life and high oral bioavailability in vivo.
Specifications

Chemical Identifiers

134470-38-5
316.332
bw b70c, bwb70c, bw-b70c, bw-b 70c, chembl86676, e-n-3-3-4-fluorophenoxy phenyl-1-r,s-methylprop-2-enyl-n-hydroxyurea, n-3-3-4-fluorophenoxy phenyl-1-methyl-2-propenyl-n-hydroxyurea, urea, n-3-3-4-fluorophenoxy phenyl-1-methyl-2-propenyl-n-hydroxy, 1-3e-4-3-4-fluorophenoxy phenyl but-3-en-2-yl-1-hydroxyurea
CHEBI:75307
CC(C=CC1=CC(=CC=C1)OC2=CC=C(C=C2)F)N(C(=O)N)O
C17H17FN2O3
UAIYNMRLUHHRMF-AATRIKPKSA-N
5353454
1-[(E)-4-[3-(4-fluorophenoxy)phenyl]but-3-en-2-yl]-1-hydroxyurea

Specifications

10mg
134470-38-5
bw b70c, bwb70c, bw-b70c, bw-b 70c, chembl86676, e-n-3-3-4-fluorophenoxy phenyl-1-r,s-methylprop-2-enyl-n-hydroxyurea, n-3-3-4-fluorophenoxy phenyl-1-methyl-2-propenyl-n-hydroxyurea, urea, n-3-3-4-fluorophenoxy phenyl-1-methyl-2-propenyl-n-hydroxy, 1-3e-4-3-4-fluorophenoxy phenyl but-3-en-2-yl-1-hydroxyurea
CC(C=CC1=CC(=CC=C1)OC2=CC=C(C=C2)F)N(C(=O)N)O
316.332
CHEBI:75307
>98%
BW-B 70C
C17H17FN2O3
UAIYNMRLUHHRMF-AATRIKPKSA-N
1-[(E)-4-[3-(4-fluorophenoxy)phenyl]but-3-en-2-yl]-1-hydroxyurea
5353454
316.33
Videos
Documents