Learn More
BW 723C86 hydrochloride, Tocris Bioscience™
5-HT2B agonist
Brand:  Tocris (Bio-Techne) 105910
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13488403
  • £133.65 / 10mg
Estimated Shipment
04-10-2018
Add to basket
Description
Description
Selective 5-HT2B receptor agonist.
Specifications
Chemical Identifiers
160521-72-2
322.851
α-methyl-5-(2-thienylmethoxy)-1H-indole-3-ethanamine hydrochloride
C16H19ClN2OS
PYJBJMIBANAOFJ-UHFFFAOYSA-N
5311036
Specifications
BW 723C86 hydrochloride
160521-72-2
C16H19ClN2OS
PYJBJMIBANAOFJ-UHFFFAOYSA-N
1-[5-(thiophen-2-ylmethoxy)-1H-indol-3-yl]propan-2-amine;hydrochloride
5311036
10mg
>99%
α-methyl-5-(2-thienylmethoxy)-1H-indole-3-ethanamine hydrochloride
CC(CC1=CNC2=C1C=C(C=C2)OCC3=CC=CS3)N.Cl
322.851
322.85
Videos