missing translation for 'onlineSavingsMsg'
Learn More

Butyrophenone, 99%, Thermo Scientific Chemicals

Catalog Number p-7081394
Click to view available options
Quantity:
25 g
50 g
250 g
This item is not returnable. View return policy
495-40-9
C10H12O
148.205
MFCD00009397
FFSAXUULYPJSKH-UHFFFAOYSA-N
butyrophenone, n-butyrophenone, 1-butanone, 1-phenyl, phenyl propyl ketone, 1-phenyl-1-butanone, propyl phenyl ketone, unii-186i11wb5b, 1-phenyl-butan-1-one, propylphenylketone
10315
1-phenylbutan-1-one
CCCC(=O)C1=CC=CC=C1
This item is not returnable. View return policy
495-40-9
C10H12O
148.205
MFCD00009397
FFSAXUULYPJSKH-UHFFFAOYSA-N
butyrophenone, n-butyrophenone, 1-butanone, 1-phenyl, phenyl propyl ketone, 1-phenyl-1-butanone, propyl phenyl ketone, unii-186i11wb5b, 1-phenyl-butan-1-one, propylphenylketone
10315
1-phenylbutan-1-one
CCCC(=O)C1=CC=CC=C1

Butyrophenone is used as intermediates of Liquid Crystals

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Butyrophenone is used as intermediates of Liquid Crystals

Solubility
Insoluble in water.

Notes
Protect from light. Store under an inert atmosphere.
TRUSTED_SUSTAINABILITY
Melting Point 11°C to 13°C
Density 1.021
Boiling Point 222°C
Flash Point 88°C (190°F)
Odor Characteristic
Refractive Index 1.5195
Quantity 25 g
Beilstein 508305
Solubility Information Insoluble in water.
Formula Weight 148.21
Percent Purity 99%
Chemical Name or Material Butyrophenone
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'