missing translation for 'onlineSavingsMsg'
Learn More

Butyraldehyde diethylacetal, 97%, Thermo Scientific Chemicals

Catalog Number 10494635
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
3658-95-5
C8H18O2
146.23
MFCD00038316
UVHXZFGCCJLFMX-UHFFFAOYSA-N
butane, 1,1-diethoxy, butyraldehyde, diethyl acetal, butyraldehyde diethyl acetal, butylaldehyde diethyl acetal, n-butyraldehyde diethyl acetal, butyraldehyde diethyl acetal, analytical standard
77225
1,1-diethoxybutane
CCCC(OCC)OCC
This item is not returnable. View return policy
3658-95-5
C8H18O2
146.23
MFCD00038316
UVHXZFGCCJLFMX-UHFFFAOYSA-N
butane, 1,1-diethoxy, butyraldehyde, diethyl acetal, butyraldehyde diethyl acetal, butylaldehyde diethyl acetal, n-butyraldehyde diethyl acetal, butyraldehyde diethyl acetal, analytical standard
77225
1,1-diethoxybutane
CCCC(OCC)OCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless
Density 0.8300g/mL
Boiling Point 143.0°C
Flash Point 30°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH2CH2CH(OCH2CH3)2
Refractive Index 1.3950 to 1.3970 (20°C, 589nm)
Quantity 5 g
Specific Gravity 0.83
Solubility Information Solubility in water: practically insoluble
Formula Weight 146.23
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Butyraldehyde diethylacetal
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Butyraldehyde diethylacetal
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'