missing translation for 'onlineSavingsMsg'
Learn More

Butyraldehyde, 99%, Thermo Scientific Chemicals

Catalog Number p-3735896
19.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
100 mL
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
123-72-8
C4H8O
72.11
MFCD00007023
ZTQSAGDEMFDKMZ-UHFFFAOYSA-N
butyraldehyde, n-butyraldehyde, butyral, 1-butanal, butyric aldehyde, butanaldehyde, butaldehyde, butalyde, butal, n-butanal
261
CHEBI:15743
butanal
CCCC=O
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
123-72-8
C4H8O
72.11
MFCD00007023
ZTQSAGDEMFDKMZ-UHFFFAOYSA-N
butyraldehyde, n-butyraldehyde, butyral, 1-butanal, butyric aldehyde, butanaldehyde, butaldehyde, butalyde, butal, n-butanal
261
CHEBI:15743
butanal
CCCC=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -96.0°C
Color Colorless
Density 0.8170g/mL
Boiling Point 75.0°C
Flash Point −12°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH2CH2CHO
Refractive Index 1.3770 to 1.3830
Quantity 100 mL
Beilstein 01, 662
Merck Index 15, 1595
Specific Gravity 0.817
Solubility Information Solubility in water: 7.1% (25°C). Other solubilities: miscible with ethanol, ether, ethyl acetate,, acetone and toluene
Viscosity 0.43 mPa.s (20°C)
Formula Weight 72.11
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Butyraldehyde
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Butyraldehyde
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'