missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Butyl stearate, tech.

Brand:  Alfa Aesar™ 036338.A3

Code : NEW

Additional Details : CAS Number : 123-95-5 Weight : 2.00000kg

Quantity 2kg

 View more versions of this product

Product Code. 11303428

  • £66.00 / 2kg


Estimated Shipment date
from Supplier 29-05-2018
Add to basket

Specifications

Chemical Identifiers

Butyl stearate
Technical Grade
340.59
1792866
123-95-5
CH3(CH2)16COO(CH2)3CH3
MFCD00026669
14,1589

Specifications

Butyl stearate
Technical Grade
CH3(CH2)16COO(CH2)3CH3
MFCD00026669
1.4430
0.861g/mL
17° to 22°C
2kg
123-95-5
Liquid
340.59
1792866
14,1589
343°C
Colorless to pale yellow
160° (320°F)
Videos
SDS