missing translation for 'onlineSavingsMsg'
Learn More

Butyl Acetate, puriss. p.a., ACS Reagent, ≥99.5% (GC), Honeywell Riedel-de Haën™

Catalog Number p-7120267
Click to view available options
Quantity:
500 mL
1 L
2.5 L
6 x 1 L
Packaging:
Glass Bottle
This item is not returnable. View return policy
123-86-4
C6H12O2
116.16
MFCD00009445
DKPFZGUDAPQIHT-UHFFFAOYSA-N
n-butyl acetate, acetic acid, butyl ester, 1-butyl acetate, butyl ethanoate, acetic acid butyl ester, n-butyl ethanoate, n-butylacetate, butylacetat, acetic acid n-butyl ester, acetate de butyle
31272
CHEBI:31328
butyl acetate
CCCCOC(C)=O
This item is not returnable. View return policy
123-86-4
C6H12O2
116.16
MFCD00009445
DKPFZGUDAPQIHT-UHFFFAOYSA-N
n-butyl acetate, acetic acid, butyl ester, 1-butyl acetate, butyl ethanoate, acetic acid butyl ester, n-butyl ethanoate, n-butylacetate, butylacetat, acetic acid n-butyl ester, acetate de butyle
31272
CHEBI:31328
butyl acetate
CCCCOC(C)=O

Puriss. p.a., ACS Reagent

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY
Melting Point -78°C
Density 0.88g/mL at 25°C
Boiling Point 126°C
Explosion Limit 0.076
Flash Point 23°C (73.4°F)
Ignition Point 790°F
Packaging Glass Bottle
Linear Formula CH3COO(CH2)3CH3
Refractive Index n20/D 1.394
Quantity 500 mL
UN Number UN1123
Beilstein 1741921
Residue after Evaporation 0 to 0.001%
Vapor Density 4 (vs air)
Formula Weight 116.16g/mol
Vapor Pressure 15mmHg (25°C), 8mmHg (20°C)
Percent Purity ≥99.5% (GC)
Chemical Name or Material Butyl acetate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Butyl acetate
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'