missing translation for 'onlineSavingsMsg'
Learn More

Butabindide oxalate, Tocris Bioscience™

CCK-inactivating serine protease inhibitor

Brand:  Tocris (Bio-Techne) 132350

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13459143

  • £665.57 / 50mg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Potent, reversible, selective and competitive inhibitor of a CCK-inactivating serine protease (tripeptidyl peptidase II) (Ki = 7 nM). Active in vivo (ID50 = 1.1 and 6.8 mg/kg i.v. for inhibition of liver and brain enzyme respectively).
Specifications

Chemical Identifiers

185213-03-0
393.44
butabindide oxalate, 2s-1-2s-2-amino-1-oxobutyl-n-butyl-2,3-dihydro-1h-indole-2-carboxamide oxalate, oxalic acid-2s-1-2s-2-aminobutanoyl-n-butyl-2,3-dihydro-1h-indole-2-carboxamide 1/1
(2S)-1-[(2S)-2-aminobutanoyl]-N-butyl-2,3-dihydroindole-2-carboxamide;oxalic acid
C19H27N3O6
KKMJFDVOXSGHBF-SLHAJLBXSA-N
56972132
CCCCNC(=O)C1CC2=CC=CC=C2N1C(=O)C(CC)N.C(=O)(C(=O)O)O

Specifications

Butabindide oxalate
185213-03-0
butabindide oxalate, 2s-1-2s-2-amino-1-oxobutyl-n-butyl-2,3-dihydro-1h-indole-2-carboxamide oxalate, oxalic acid-2s-1-2s-2-aminobutanoyl-n-butyl-2,3-dihydro-1h-indole-2-carboxamide 1/1
CCCCNC(=O)C1CC2=CC=CC=C2N1C(=O)C(CC)N.C(=O)(C(=O)O)O
393.44
393.44
50mg
C19H27N3O6
KKMJFDVOXSGHBF-SLHAJLBXSA-N
(2S)-1-[(2S)-2-aminobutanoyl]-N-butyl-2,3-dihydroindole-2-carboxamide;oxalic acid
56972132
>99%
Videos